Learn More
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 038714.K2
Code : NEW
Additional Details : CAS Number : 1336-21-6 Weight : 0.01000kg
Description
Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
1 L | |
Odorless | |
23 hPa (17mm Hg) at 20°C | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 |
1336-21-6 | |
Liquid | |
H5NO | |
MFCD00066650 | |
Miscible with water. | |
N.O | |
35.05 | |
CHEBI:18219 |
RUO – Research Use Only