missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 4.01, METTLER TOLEDO™

Product Code. 10711765
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
1500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10711765 250 mL 1500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10711765 Supplier Mettler Toledo Online Supplier No. 51350042
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the perfect product for regulated industries.

TRUSTED_SUSTAINABILITY

Specifications

Color Light Red
Description Certified buffer pH 4.01, 6 x 250mL
pH 4.01
Physical Form Liquid
Quantity 250 mL
Solution Type pH Buffer
Odor None
Packaging 6x 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.