missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use

Product Code. 10727913 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10727913 500 mL 500mL
10270023 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10727913 Supplier Thermo Scientific Acros Supplier No. 383835000
  • ONLINE EXCLUSIVE PRICE
    37.30 €
    36.20 € / 500mL
    Save 1.10 €
    2% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 877-24-7 | C8H5KO4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 4
CAS 877-24-7, 7732-18-5, 7487-94-7
Concentration 1.0% (Potassium hydrogen phthalate)
pH 4.0
Physical Form Liquid
Quantity 500 mL
Molecular Formula C8H5KO4
Odor Odorless
MDL Number MFCD00217672
Solubility Information Solubility in water: miscible.
Formula Weight 204.22
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.