missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Color coded and packaged for convenience
- Colored yellow for easy identification of pH value
- Supplied in Bag in Box bulk cubitainer with integrated tap for easy dispensing or Twin Neck bottles that feature an integrated calibrating chamber
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
Specifications
| Color | Yellow |
| pH | 7.00 ±0.01 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Odor | Characteristic |
| Packaging | Twin Neck bottle with integrated calibration chamber |
| Traceability to NIST | Yes |
| Solubility Information | Fully miscible in water |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
