missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 7, Phosphate buffer

Product Code. 10317531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10317531 500 mL 500mL
10448273 100 mL 100mL
10114680 1 L 1 L
11906971 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10317531 Supplier Thermo Scientific Acros Supplier No. 258595000
  • ONLINE EXCLUSIVE PRICE
    39.35 €
    38.20 € / 500mL
    Save 1.15 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7558-79-4 | H2KO4P

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Buffer solution pH 7
CAS 7558-79-4, 7732-18-5
Density 1.0030g/mL
Quantity 500 mL
Molecular Formula H2KO4P
MDL Number MFCD03097944
Packaging Plastic bottle
Specific Gravity 1.003
Merck Index 14, 7659
Vapor Pressure 14mmHg at 20°C
Solubility Information Solubility in water: soluble.
Formula Weight 136.09
Mostrar más Mostrar menos
What is the concentration of the Buffer solution pH 7, Phosphate buffer?

The Buffer solution pH 7, Phosphate buffer has a concentration of 0.1 M.

What is the composition of the Buffer solution pH 7, Phosphate buffer?

The composition of the Buffer solution pH 7, Phosphate buffer is 3.54 g potassium dihydrogen phosphate and 14.7 g di-sodium hydrogen phosphate per liter.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.