missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, Tetraoxalate Potassium, pH 1.679, Fisher Chemical™

Product Code. 12970654 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12970654 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12970654 Supplier Fisher Chemical Supplier No. SOL 013 1L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 6100-20-5
pH 1.679
Quantity 1000 mL
Molecular Formula C4H3KO8
MDL Number MFCD00150443,MFCD00150443
Packaging Bottle
InChI Key GANDVAJEIJXBQJ-UHFFFAOYSA-M
SMILES [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
IUPAC Name potassium;hydron;oxalate;dihydrate
Molecular Weight (g/mol) 218.16
PubChem CID 131698588
Chemical Name or Material Buffer Solution, Potassium Tetraoxalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.