missing translation for 'onlineSavingsMsg'
Learn More

Buffer solutions, BAKER ANALYZED™, J.T. Baker™

Product Code. p-7220849 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
1 L
Chemical Name or Material:
Borate
Phosphate
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 13538850

Brand: JT Baker 5609.0500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability

  • You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control
  • Contain 0.01% Sodium benzoate
  • Certifications: This buffer is traceable to NIST Standard reference buffer

Specifications

Certificate of Analysis Traceable to NIST Standard reference buffer
pH 10.00
Quantity 500 mL
Packaging Plastic bottle
Chemical Name or Material Borate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.