missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffered Peptone Water (Bottled)

Product Code. 11962632
Change view
Click to view available options
Fill Weight:
225 to 229 g
225.0g - 229.0g
90g + 1.5g
Packaging Type:
10 bottles with screw cap closures in a box
10 wide-necked pots with screw cap closure in a box
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Packaging Type unitSize
11962632 90g + 1.5g 10 bottles with screw cap closures in a box Pack of 10
13255469 225.0g - 229.0g 10 bottles with screw cap closures in a box Pack of 10
10701104 225 to 229 g 10 wide-necked pots with screw cap closure in a box Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11962632 Supplier Thermo Scientific™ Supplier No. BO0201Z

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Pre-enrich food samples prior to the isolation of Salmonella species using these ready-prepared media bottles.

Pre-enrich food samples prior to the isolation of Salmonella species with Thermo Scientific™ Oxoid™ Buffered Peptone Water ready-prepared bottled medium. Buffered Peptone Water is a non-selective pre-enrichment medium.

Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.

  • Convenient: the pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.
  • Aids recovery of damaged Salmonella cells.

The test sample is added to Buffered Peptone Water at a ratio of 1:10 and incubated at 37° ± 1°C for 18 hours + 2 hours.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, Clear
Content And Storage 2 to 25°C
Description Buffered Peptone Water (Bottled)
Format Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 90g + 1.5g
Packaging Type 10 bottles with screw cap closures in a box
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.