missing translation for 'onlineSavingsMsg'
Learn More

Bufuralol Hydrochloride, TRC

Product Code. 30010373
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010373 1 mg 1mg
30011183 25 mg 25mg
30010524 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010373 Supplier Toronto Research Chemicals Supplier No. TRCB6895401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60398-91-6
Molecular Formula C16 H23 N O2 . Cl H
Molecular Weight (g/mol) 297.82
IUPAC Name 2-(tert-butylamino)-1-(7-ethyl-1-benzofuran-2-yl)ethanol;hydrochloride
SMILES Cl.CCc1cccc2cc(oc12)C(O)CNC(C)(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H23NO2.ClH/c1-5-11-7-6-8-12-9-14(19-15(11)12)13(18)10-17-16(2,3)4;/h6-9,13,17-18H,5,10H2,1-4H3;1H
Formula Weight 297.1496
Percent Purity >95%
Chemical Name or Material Bufuralol, Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.