missing translation for 'onlineSavingsMsg'
Learn More

Bumetanide, 98+%

Product Code. 15454709
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454709 5 g 5g
15444709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454709 Supplier Thermo Scientific Alfa Aesar Supplier No. J62302.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of the sodium-potassium-chloride cotransporter | CAS: 28395-03-1 | C17H20N2O5S | 364.416 g/mol

Bumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1

Solubility
Soluble in ethanol (10 mg/ml), DMSO (25 mg/ml), acetone, benzene, methanol, propylene glycol, and water (<1mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28395-03-1
Molecular Formula C17H20N2O5S
Molecular Weight (g/mol) 364.416
MDL Number MFCD00078949
InChI Key MAEIEVLCKWDQJH-UHFFFAOYSA-N
Synonym bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum
PubChem CID 2471
ChEBI CHEBI:3213
IUPAC Name 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid
SMILES CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2

Specifications

Melting Point 230°C to 231°C
Color White
Quantity 5 g
Merck Index 14,1484
Solubility Information Soluble in ethanol (10mg/ml),DMSO (25mg/ml),acetone,benzene,methanol,propylene glycol,and water (<1mg/ml).
Formula Weight 364.4
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Bumetanide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.