Learn More
Bumetanide, 98+%, Thermo Scientific™
Inhibitor of the sodium-potassium-chloride cotransporter
133.00 € - 488.00 €
Chemical Identifiers
| CAS | 28395-03-1 |
|---|---|
| Molecular Formula | C17H20N2O5S |
| Molecular Weight (g/mol) | 364.416 |
| MDL Number | MFCD00078949 |
| InChI Key | MAEIEVLCKWDQJH-UHFFFAOYSA-N |
| Synonym | bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum |
| PubChem CID | 2471 |
| ChEBI | CHEBI:3213 |
| IUPAC Name | 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid |
| SMILES | CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15444709
|
Thermo Scientific Alfa Aesar
J62302.03 |
1g |
136.00 € 133.00 € / 1g Save 3.00 € 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15454709
|
Thermo Scientific Alfa Aesar
J62302.06 |
5g |
498.00 € 488.00 € / 5g Save 10.00 € 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsBumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1
Solubility
Soluble in ethanol (10 mg/ml), DMSO (25 mg/ml), acetone, benzene, methanol, propylene glycol, and water (<1mg/mL).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids.
Chemical Identifiers
| 28395-03-1 | |
| 364.416 | |
| MAEIEVLCKWDQJH-UHFFFAOYSA-N | |
| 2471 | |
| 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid |
| C17H20N2O5S | |
| MFCD00078949 | |
| bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum | |
| CHEBI:3213 | |
| CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2 |
Specifications
| 28395-03-1 | |
| MFCD00078949 | |
| bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum | |
| MAEIEVLCKWDQJH-UHFFFAOYSA-N | |
| 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid | |
| 2471 | |
| 364.4 | |
| Bumetanide |
| C17H20N2O5S | |
| 14,1484 | |
| Soluble in ethanol (10mg/ml),DMSO (25mg/ml),acetone,benzene,methanol,propylene glycol,and water (<1mg/ml). | |
| CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2 | |
| 364.416 | |
| CHEBI:3213 | |
| ≥98% |
Safety and Handling
missing translation for 'einecsNumber' : 249-004-6
missing translation for 'rtecsNumber' : DG4910000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.