missing translation for 'onlineSavingsMsg'
Learn More

Bumetanide, TRC

Product Code. 30136436
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136436 100 mg 100mg
30133690 1 g 1g
17998524 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136436 Supplier Toronto Research Chemicals Supplier No. TRCB689550100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28395-03-1
Molecular Formula C17 H20 N2 O5 S
Molecular Weight (g/mol) 364.42
Synonym Bumetanide, 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoic acid, Benzoic acid, 3-(aminosulfonyl)-5-(butylamino)-4-phenoxy-, Benzoic acid, 3-(butylamino)-4-phenoxy-5-sulfamoyl- (8CI), 3-(Aminosulfonyl)-5-(butylamino)-4-phenoxybenzoic acid, 3-(Butylamino)-4-phenoxy-5-sulfamoylbenzoic acid, 3-n-Butylamino-4-phenoxy-5-sulfamoylbenzoic acid, Bufenox, Bumetanide, Bumethanide, Bumex, Burinex, Edemex, Fontego, Fordiuran, Lixil, Lunetoron, PF 1593, Ro 10-6338
IUPAC Name 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid
SMILES CCCCNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H20N2O5S/c1-2-3-9-19-14-10-12(17(20)21)11-15(25(18,22)23)16(14)24-13-7-5-4-6-8-13/h4-8,10-11,19H,2-3,9H2,1H3,(H,20,21)(H2,18,22,23)
Formula Weight 364.1093
Percent Purity >95%
Chemical Name or Material Bumetanide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.