missing translation for 'onlineSavingsMsg'
Learn More

Butachlor (>90%), TRC

Product Code. 30137192
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137192 10 g 10g
17943435 1 g 1g
17933865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137192 Supplier Toronto Research Chemicals Supplier No. TRCB68992510G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23184-66-9
Molecular Formula C17 H26 Cl N O2
Molecular Weight (g/mol) 311.85
Synonym Acetanilide, N-(butoxymethyl)-2-chloro-2',6'-diethyl- (8CI), N-(Butoxymethyl)-2-chloro-N-(2,6-diethylphenyl)acetamide, 2-Chloro-2',6'-diethyl-N-(butoxymethyl)acetanilide, 2-Chloro-2',6'-diethyl-N-(butoxymethyl)acetoanilide, 2',6'-Diethyl-N-butoxymethyl-2-chloroacetanilide, 2',6'-Diethyl-N-butoxymethyl-α-chloroacetanilide, Bilchlor, Butachlor, Butaclor, CP 53619, Delchlor, Delchlor 5G, Dingcaoan, Hiltachlor, Machet, Machete, Machete (herbicide), Machette, N-(Butoxymethyl)-2-chloro-2',6'-diethylacetanilide, NSC 221683
IUPAC Name N-(butoxymethyl)-2-chloro-N-(2,6-diethylphenyl)acetamide
SMILES CCCCOCN(C(=O)CCl)c1c(CC)cccc1CC

Specifications

Analyte or Component Names Butachlor
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H26ClNO2/c1-4-7-11-21-13-19(16(20)12-18)17-14(5-2)9-8-10-15(17)6-3/h8-10H,4-7,11-13H2,1-3H3
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 g
Formula Weight 311.1652
Percent Purity >95%
Chemical Name or Material Butachlor
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.