missing translation for 'onlineSavingsMsg'
Learn More

Butyl 4-Hydroxybenzoate (Butyl Paraben), TRC

Product Code. 30011826
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011826 10 g 10g
30136472 5 g 5g
17912705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011826 Supplier Toronto Research Chemicals Supplier No. TRCB69360010G
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-26-8
Molecular Formula C11 H14 O3
Molecular Weight (g/mol) 194.23
Synonym Butyl 4-Hydroxybenzoate, Butyl Parahydroxybenzoate, Ethyl Parahydroxybenzoate Imp. D (EP), Propyl Parahydroxybenzoate Imp. D (EP), Methyl Parahydroxybenzoate Imp. D (EP), 4-(Butoxycarbonyl)phenol, 4-Hydroxybenzoic acid butyl ester, Aseptoform Butyl, Butoben, Butyl 4-hydroxybenzoate, Butyl Butex, Butyl Tegosept, Butyl chemosept, Butyl p-hydroxybenzoate, Butyl parabens, Butyl parasept, Butylparaben, Mekkings B, Mekkins B, NSC 13164, NSC 8475, Nipabutyl, Preserval B, SPF, Solbrol B, Tegosept B, Tegosept Butyl, n-Butyl 4-hydroxybenzoate, n-Butyl p-hydroxybenzoate, n-Butylparaben, p-Hydroxybenzoic acid butyl ester
IUPAC Name butyl 4-hydroxybenzoate
SMILES CCCCOC(=O)c1ccc(O)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3
Formula Weight 194.0943
Percent Purity >95%
Chemical Name or Material Butyl Parahydroxybenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.