missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxyanisole 500 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30316561
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316561 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316561 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010379ME
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl-4-hydroxyanisole (mixture of 2- and 3-isomer)
CAS 25013-16-5
Analyte or Component Names tert-Butyl-4-hydroxyanisole (mixture of 2- and 3-isomer)
Shipping Condition Room Temperature
Molecular Formula 2 C11 H16 O2
Concentration 500 μg/mL
Certifications/Compliance ISO 17034
Synonym tert-Butyl-4-methoxyphenol, (1,1-Dimethylethyl)-4-methoxyphenol, 2(3)-tert-Butyl-4-hydroxyanisole, Antioxyne B, BHA, BHA (antioxidant), BOA, BOA (antioxidant), Butylated hydroxyanisole, Butylhydroxyanisole, E 320, Embanox, Grindox BHA, Protex, Sustane 1F, Tenox BHA, tert-Butyl-4-hydroxyanisole, tert-Butyl-p-Hydroxyanisol, tert-Butyl-p-hydroxyanisole, tert-Butylhydroxyanisole, Butylated Hydroxyanisole
InChI Formula InChI=1S/2C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12;1-11(2,3)9-7-8(12)5-6-10(9)13-4/h2*5-7,12H,1-4H3
SMILES COc1ccc(O)c(c1)C(C)(C)C.COc2ccc(O)cc2C(C)(C)C
IUPAC Name 2-tert-butyl-4-methoxyphenol;3-tert-butyl-4-methoxyphenol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 360.49
Quantity 1 mL
Formula Weight 360.2301 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.