missing translation for 'onlineSavingsMsg'
Learn More

Butylated hydroxyanisole, 96%

Product Code. 10665282 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665282 500 g Plastic bottle 500g
10551291 100 g Plastic bottle 100g
10648324 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10665282 Supplier Thermo Scientific Acros Supplier No. 235235000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 25013-16-5 | C11H16O2 | 180.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.24
MDL Number MFCD01779059
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
Synonym 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
PubChem CID 8456
ChEBI CHEBI:76358
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=C(C=CC(=C1)OC)O

Specifications

Melting Point 48°C to 55°C
Color White to Yellow
Boiling Point 268°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Merck Index 15,1549
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,propylene glycol,,chlorogorm,ether and acetone,slightly soluble in benzene
Formula Weight 180.24
Percent Purity 96%
Physical Form Crystalline Flakes or Crystals
Chemical Name or Material Butylated hydroxyanisole, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butylated hydroxyanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.