missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxytoluene Crystalline MP Biomedicals

Product Code. 11348769 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348769 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11348769 Supplier MP Biomedicals Supplier No. 0210116280
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 128-37-0
Density 7.6 (air = 1)
Molecular Formula C15H24O
Quantity 100 g
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
IUPAC Name 2,6-ditert-butyl-4-methylphenol
Molecular Weight (g/mol) 220.356
PubChem CID 31404
ChEBI CHEBI:34247
Formula Weight 220.34
Physical Form Crystalline
Chemical Name or Material Butylated Hydroxytoluene
Show More Show Less
compliance-icons
Product Identifier
  • Butylated Hydroxytoluene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Butylated Hydroxytoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.