missing translation for 'onlineSavingsMsg'
Learn More

Butylhydroxyanisole, MedChemExpress

Product Code. 16462309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462309 Supplier Medchem Express Supplier No. HYB1066
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Butylhydroxyanisole (Butylated hydroxyanisole) is an antioxidant used as a food additive preservative. Butylhydroxyanisole mediates liver toxicity, retardation in reproductive organ development and learning, and sleep deficit. Butylhydroxyanisole exerts neurotoxic effects and leads to disruption of the brain and nerve development. Butylhydroxyanisole is a ferroptosis inducer.

Butylhydroxyanisole (Butylated hydroxyanisole) is an antioxidant used as a food additive preservative. Butylhydroxyanisole mediates liver toxicity, retardation in reproductive organ development and learning, and sleep deficit. Butylhydroxyanisole exerts neurotoxic effects and leads to disruption of the brain and nerve development. Butylhydroxyanisole is a ferroptosis inducer.

Specifications

Chemical Name or Material Butylhydroxyanisole
CAS 25013-16-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H16O2
Quantity 1 g
Synonym Butylated hydroxyanisole BHA E320
Solubility Information DMSO : ≥ 100 mg/mL (554.82 mM) ∣H2O : 1 mg/mL (5.55 mM; ultrasonic and warming and heat to 60°C)
SMILES OC1=CC=C(OC)C=C1C(C)(C)C.OC2=CC=C(OC)C(C(C)(C)C)=C2
Molecular Weight (g/mol) 180.24
Formula Weight 180.24
Percent Purity 99.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.