missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyrylcholinesterase/BCHE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98274-100ul
This item is not returnable.
View return policy
Description
Butyrylcholinesterase/BCHE Polyclonal antibody specifically detects Butyrylcholinesterase/BCHE in Mouse samples. It is validated for ELISA
Specifications
| Butyrylcholinesterase/BCHE | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Acylcholine acylhydrolase, Butyrylcholine esterase, butyrylcholinesterase, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase | |
| Produced in rabbits immunized with purified, recombinant Mouse Butyrylcholinesterase/BCHE (Accession#: NP_033868.3; Met 1-Leu 603) | |
| 100 μg | |
| Alzheimers Research, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 590 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction