missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BXDC2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148793
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Required for biogenesis of the 60S ribosomal subunit.
Specifications
| BXDC2 | |
| Polyclonal | |
| Unconjugated | |
| Brix1 | |
| 1110064N10Rik; BRIX; brix domain containing 2; brix domain-containing protein 2; Brix1; BRX1, biogenesis of ribosomes; BRX1, biogenesis of ribosomes, homolog; BRX1, biogenesis of ribosomes, homolog (S. cerevisiae); BXDC2; C76935; RGD1308508; ribosome biogenesis protein Brix; ribosome biogenesis protein BRX1 homolog | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 55299 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8TDN6 | |
| Brix1 | |
| E. coli-derived recombinant human BRIX, Ser131-Lys353. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction