missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Fos Antibody (2H2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50037B
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
c-Fos Monoclonal antibody specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Gene Knock-Out
Tekniske data
| c-Fos | |
| Monoclonal | |
| Biotin | |
| activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos | |
| This c-Fos Antibody (2H2) was developed against full length recombinant human c-Fos protein expressed in and purified from E. coli. [UniProt# P01100] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors | |
| 2353 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence, Gene Knock-Out | |
| 2H2 | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion