missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Maf Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-24551
This item is not returnable.
View return policy
Description
c-Maf Polyclonal specifically detects c-Maf in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| c-Maf | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, c-MAF, c-maf proto-oncogene, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4094 | |
| Human, Mouse, Rat, Bovine | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 2-4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml | |
| O75444 | |
| MAF | |
| A synthetic peptide corresponding to amino acids 75-110 of human c-maf was used as the immunogen, GenBank no. NP_001026974.1 . | |
| 0.1 mg | |
| Primary | |
| 100% homologous in human isoforms CRA_a, CRA_b, and CRA_c). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction