missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Maf Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88072
This item is not returnable.
View return policy
Description
c-Maf Polyclonal antibody specifically detects c-Maf in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| c-Maf | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, c-MAF, c-maf proto-oncogene, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian) | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MASELAMSNSDLPTSPLAMEYVNDFDLMKFEVKKEPVETDRIISQCGRLIAGGSLSSTPMSTPCSSVPPSPSFSAPSPGSGSEQKAHLEDYYWMTGYPQQLNPEALGFSPED | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Immunology | |
| 4094 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction