missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ c-MAF Monoclonal Antibody (883926)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561928
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
c-maf is the cellular counterpart of oncogenic v-maf that belongs to the family of basic region leucine zipper domain transcription factors. The leucine-zipper domain is involved in the interaction with LRPICD. There are two forms of human c-maf mRNA, c-maf-long and c-maf-short. It is identified in the genome of the acute transforming avian retrovirus AS42. c-maf targets are IL-4 in Th2 cells, the crystalline genes in lens fiber cells, insulin gene in islet, p53 and L7 where it exerts its transcriptional role through binding to a Maf recognition element (MARE). It regulates Th2 differentiation and lineage-specific hematopoiesis. c-maf is a transcription factor for IL-10 gene expression in LPS-activated macrophages. Chromosomal aberration involving maf is found in some forms of multiple myeloma. It is expressed in myeloma cell lines and resting monocytes/macrophages.
Specifications
| c-MAF | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O75444 | |
| MAF | |
| E. coli-derived recombinant human c-Maf, Met1-Phe372. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 883926 | |
| Unconjugated | |
| MAF | |
| 2810401A20Rik; A230108G15Rik; Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene; avian musculoaponeurotic fibrosarcoma (v-maf) AS42 oncogene homolog; AW047063; AYGRP; basic domain leucine zipper transcription factor; CCA4; cMaf; C-Maf; c-Maf long form; c-maf proto-oncogene; CTRCT21; fl13d12; hMafA; LOW QUALITY PROTEIN: transcription factor Maf; LOW QUALITY PROTEIN: transcription factor MafA; maf; MAF bZIP transcription factor; MAF bZIP transcription factor A; maf protein; Maf2; MAFA; Mafa homolog; MGC71685; pancreatic beta-cell-specific transcriptional activator; Proto-oncogene c-Maf; RGD1562627; RIPE3b1; RIPE3b1 factor; T lymphocyte c-maf long form; transcription factor Maf; Transcription factor Maf-2; transcription factor MafA; transcription factor RIPE3b1; v-maf avian musculoaponeurotic fibrosarcoma oncogene A; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog A; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog a (paralog a); v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog; V-maf musculoaponeurotic fibrosarcoma oncogene homolog A; wu:fl13d12; Z-cmaf | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4094 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction