missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (MYC/7682R), Novus Biologicals™
Rabbit Monoclonal Antibody
287.00 € - 519.00 €
Specifications
| Antigen | c-Myc |
|---|---|
| Clone | MYC/7682R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640075
|
Novus Biologicals
NBP3-20661-20ug |
20 μg |
287.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672515
|
Novus Biologicals
NBP3-20661-100ug |
100 μg |
519.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Myc Monoclonal antibody specifically detects c-Myc in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| c-Myc | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 10mM PBS with 0.05% BSA | |
| 4609 | |
| IgG κ | |
| Protein A or G purified |
| MYC/7682R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| Recombinant fragment (around aa1-200) of human c-Myc protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title