Learn More
C-Reactive Protein/CRP Antibody (C7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB600-1333H
Product Code. 18256992
  • 474.00 € / 0.20mL
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay, Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C-Reactive Protein/CRP
Monoclonal
HRP
PBS with No Preservative
CRP
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human CRP.
0.2 mL
Cancer, Cytokine Research, Immunology, Innate Immunity
1401
Human
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
C7
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay, Sandwich ELISA
C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895
Mouse
Protein A purified
RUO
Primary
Human CRP
Store at 4C in the dark.
IgG1