missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1 inhibitor Recombinant Rabbit Monoclonal Antibody (18), HRP, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
163.00 € - 510.00 €
Specifications
| Antigen | C1 inhibitor |
|---|---|
| Clone | 18 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019845
![]() |
Invitrogen™
MA552269 |
100 μg |
510.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019948
|
Invitrogen™
MA552270 |
20 μg |
163.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C1 inhibitor Recombinant Monoclonal antibody specifically detects C1 inhibitor in Human samples. It is validated for ELISASpecifications
| C1 inhibitor | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1Inh; C1-inhibiting factor; C1nh; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH | |
| Recombinant Human SerpinG1 protein (Asn 23-Ala500). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| SERPING1 |
| 18 | |
| ELISA | |
| HRP | |
| Rabbit | |
| Human | |
| P05155 | |
| 710 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title