missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf119 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-82164
This item is not returnable.
View return policy
Description
C14orf119 Polyclonal specifically detects C14orf119 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| C14orf119 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| chromosome 14 open reading frame 119, FLJ20671, FLJ21141, MGC74723, My028 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55017 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C14ORF119 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SLLPSVPHNTNPSPPLMSYITSQEMKCILHWFANWSGPQRERFLEDLVAKAVPEKLQPLLDSLEQLSVSGADRPPSIFECQLHLWDQWFRGWAEQERNEFVRQLEFSEPDFVAKFYQ | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction