missing translation for 'onlineSavingsMsg'
Learn More

C17orf37 Monoclonal Antibody (OTI1D7), TrueMAB™, OriGene

Product Code. 17561962
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17561962 30 μL 30µL
17541962 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17561962 Supplier Origene Technologies Supplier No. TA504583S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C17orf37 Monoclonal antibody specifically detects C17orf37 in Human samples. It is validated for Western Blot

Specifications

Antigen C17orf37
Applications Western Blot
Classification Monoclonal
Clone OTI1D7
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CYB5R1
Gene Accession No. Q9UHQ9
Gene Alias 1810046J19Rik; AI463380; C16H17orf37; C17orf37; C35; HBV XAg-transactivated protein 4; HBV X-transactivated gene 4 protein; MGC14832; MIEN1; Migration and invasion enhancer 1; ORB3; protein C17orf37; protein C17orf37 homolog; Protein C35; RDX12; RGD1306682; XTP4
Host Species Mouse
Immunogen Full length human recombinant protein of human CYB5R1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51706
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.