missing translation for 'onlineSavingsMsg'
Learn More

C20orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16836314 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16836314 20 μL 20µL
16826314 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16836314 Supplier Proteintech Supplier No. 166331AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Product No. 16633-1-AP is a rabbit polyclonal antibody raised against the full-length of human C20orf43. The MW of this protein is 40 kDa, and this antibody specifically recognizes the 40 kDa protein.

C20orf43 protein function remains unknown.

Specifications

Antigen C20orf43
Applications Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RTFDC1
Gene Accession No. Q3T1J8, Q99K95, Q9BY42
Gene Alias C20orf43, CDA05, CDAO5, HSPC164, HSPC169, RTF2, SHUJUN 3, UPF0549 protein C20orf43
Gene Symbols Rtfdc1
Host Species Rabbit
Immunogen C20orf43 Fusion Protein Ag9972
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296410, 51507, 66404
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.