missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C21orf62 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
220.00 € - 326.00 €
Specifications
| Antigen | C21orf62 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671838
|
Novus Biologicals
NBP3-06003-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605998
|
Novus Biologicals
NBP3-06003-50ul |
50 μg |
220.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C21orf62 Polyclonal antibody specifically detects C21orf62 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| C21orf62 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| B37, B37PRED81, C21orf120, chromosome 21 open reading frame 120, chromosome 21 open reading frame 62, hypothetical protein LOC56245, PRED81 | |
| Produced in rabbits immunized with E. coli-derived Human C21orf62 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 56245 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title