missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C22orf15 Recombinant Mouse Monoclonal Antibody (K9N021_14D4)
GREENER_CHOICE

Product Code. 30828554
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828554 100 μL
30830373 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828554 Supplier Invitrogen™ Supplier No. MA564446

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: CSVLVNTSCR LVNLTAHLRQ KAGLPPDATI ALLAEDGNLV SLEEDLKEGA SRAQTMGNSL LKERAIYVLV RIIKGEDMAS TRYESLLENL DDHYPELAEE LRRLSGLSSV Highest antigen sequence identity to the following orthologs: Pig - 68%, Cynomolgus monkey - 94%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C22orf15
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K9N021_14D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene C22orf15
Gene Accession No. Q8WYQ4
Gene Alias C22orf15; chromosome 22 open reading frame 15; N27C7-3; protein N27C7-3; Uncharacterized protein C22orf15
Gene Symbols C22orf15
Host Species Mouse
Immunogen Recombinant human C22orf15 (aa 11-120).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150248
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.