missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2CD5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92738-0.02ml
This item is not returnable.
View return policy
Description
C2CD5 Polyclonal antibody specifically detects C2CD5 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| C2CD5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp779N2044, hypothetical protein LOC9847, KIAA0528 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 732-1051 of human C2CD5 (NP_001273103.1). RLSSLNLTNQALNKNFNDLCENLLKSLYFKLRSMIPCCLCHVNFTVSLPEDELIQVTVTAVAITFDKNQALQTTKTPVEKSLQRASTDNEELLQFPLELCSDSLPSHPFPPAKEHLESASSNSGIPAAQRATSVDYSSFADRCSSWIELIKLKAQTIRRGSIKTTMTVEKASPVGDGNFRNRSAPPCANSTVGVVKMTPLSFIPGAKITKYLGIINMFFIRETTSLREEGGVSGFLHAFIAEVFAMVRAHVAALGGNAVVSYIMKQCVFMENPNKNQAQCLINVSGDAVVFVRESDLEVVSSQQPTTNCQSSCTEGEVTT | |
| 0.02 mL | |
| Cell Biology, Signal Transduction | |
| 9847 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction