missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf72 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | C2orf72 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18471561
|
Novus Biologicals
NBP1-93551-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18061347
|
Novus Biologicals
NBP1-93551 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
C2orf72 Polyclonal specifically detects C2orf72 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
| C2orf72 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 257407 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GQPVEGAWERPGLPGLLACFSWGPWSRRKNQDVAACRSSAQEDFQEPEEELPLTAIFPNGDCDDLGRGSKACDGVVHTPAEP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| chromosome 2 open reading frame 72, hypothetical protein LOC257407 | |
| C2orf72 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts