missing translation for 'onlineSavingsMsg'
Learn More

C3 Ceramide, TRC

Product Code. 30351590
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351590 5 mg 5mg
30355786 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351590 Supplier Toronto Research Chemicals Supplier No. TRCC2625205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362678-52-2
Molecular Formula C21H41NO3
Molecular Weight (g/mol) 355.56
Synonym N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]propanamide, N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]propanamide
IUPAC Name N-((2S,3R,E)-1,3-dihydroxyoctadec-4-en-2-yl)propionamide
SMILES OC[C@H](NC(CC)=O)[C@H](O)/C=C/CCCCCCCCCCCCC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H41NO3/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-20(24)19(18-23)22-21(25)4-2/h16-17,19-20,23-24H,3-15,18H2,1-2H3,(H,22,25)/b17-16+/t19-,20+/m0/s1
Formula Weight 355.31
Chemical Name or Material C3 Ceramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.