missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CA125/MUC16 Antibody (sofituzumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28082-100ug
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CA125/MUC16 | |
| Monoclonal | |
| Unconjugated | |
| Q8WXI7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| sofituzumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| MUC16 | |
| 100 μg | |
| Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
| 94025 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction