missing translation for 'onlineSavingsMsg'
Learn More

CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-1, Mouse Monoclonal Antibody.

124.44 € - 385.00 €

Specifications

Antigen CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) Ab-1
Clone 121SLE
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12671247
View Documents
Epredia
MS-379-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12641257
View Documents
Epredia
MS-379-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12601257
View Documents
Epredia
MS-379-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12691247
View Documents
Epredia
MS-379-P1
500 μL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
286.00 €
239.79 € / 500µL
Save 46.21 €
16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12681247
View Documents
Epredia
MS-379-P
1 mL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
407.00 €
342.01 € / 1mL
Save 64.99 €
15% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12611257
View Documents
Epredia
MS-379-R7
7 mL Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Mucin glycoprotein is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas, including adenocarcinomas of the stomach, intestine, and pancreas. Preoperative elevated CA19-9 levels in patients with stage I pancreatic carcinoma decrease to normal values following surgery. When used serially, CA19-9 can predict recurrence of disease prior to radiographic or clinical findings.

Host Species: Mouse

Clone: 121SLE

Isotype: IgM

Species Reactivity: Human. Does not react with rat. Others not tested.

Immunogen: Mucins from ovarian cyst

Molecular Weight: >400kDa

Positive Control: Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgM
121SLE
Monoclonal
Mouse
Oncogenesis
Mucins from ovarian cyst
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.