missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CA2 (Human) Recombinant Protein
Human CA2 partial ORF (NP_000058.1, 191 a.a. - 260 a.a.) recombinant protein with GST tag at N-terminal.
Brand: Abnova™ H00000760-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
- Sequence: WTYPGSLTTPPLLECVTWIVLKEPISVSSEQVLKFRKLNFNGEGEPEELMVDNWRPAQPLKNRQIKASFK
Specifications
NP_000058.1 | |
carbonic anhydrase II | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CA2 | |
GST |
760 | |
Wheat germ expression system | |
10 μg | |
CA-II, CAII, Car2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |