Learn More
Invitrogen™ CAB39 Recombinant Rabbit Monoclonal Antibody (24GB6950)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556995
 View more versions of this product
Product Code. 30276627
  • 500.00 € / 100µL
Description
CAB39 Recombinant Monoclonal Antibody for Western Blot
Peutz-Jeghers Syndrome (PJS) is a rare hereditary disease characterized by melanocytic macules of the lips, gastrointestinal hamartomatous polyps and an increased risk for many classes of cancer. The serine/threonine kinase LKB1 (also designated STK11) has been identified as the gene mutated in PJS. LKB1 activity increases upon the binding of a regulatory complex consisting of the STE20-related adaptor-alpha (STRAD alpha) pseudo kinase and the calcium binding protein 39 (MO25 alpha). STRAD and MO25 determine the subcellular localization of LKB1 by initiating its translocation from the nucleus to the cytoplasm, thus regulating the tumor suppressor activity of LKB1. The LKB1/STRAD/MO25 complex acts as an AMP-activated protein kinase kinase (AMPKK).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CAB39
Recombinant Monoclonal
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q06138, Q9Y376
CAB39
A synthesized peptide derived from human CAB39/MO25 (aa 323-341).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6950
Unconjugated
CAB39
AA408805; AA960512; C78372; Cab39; calcium binding protein 39; calcium binding protein, 39 kDa; calcium-binding protein 39; CGI-66; MO25; MO25alpha; Protein Mo25
Rabbit
Affinity chromatography
RUO
12283, 301574, 51719
-20°C