missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNA1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30941-25ug
This item is not returnable.
View return policy
Description
CACNA1A Polyclonal antibody specifically detects CACNA1A in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| CACNA1A | |
| Polyclonal | |
| Western Blot : 1:400, Immunohistochemistry | |
| APCA, BI, brain calcium channel 1, Brain calcium channel I, CACH4, CACN3, CACNL1A4CAV2.1, calcium channel, L type, alpha-1 polypeptide, Calcium channel, L type, alpha-1 polypeptide isoform 4, calcium channel, voltage-dependent, P/Q type, alpha 1A subunit, Cav2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6, voltage-dependent P/Q-type calcium channel subunit alpha-1A, Voltage-gated calcium channel subunit alpha Cav2.1 | |
| Synthetic peptide derived from the rat CACNA1A conjugated to KLH | |
| 25 μg | |
| Signal Transduction | |
| 773 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction