Learn More
CACNA1E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-43404
Product Code. 30279617
  • 310.00 € / 100µL
Description
CACNA1E Polyclonal antibody specifically detects CACNA1E in Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Serum,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CACNA1E
Polyclonal
Unconjugated
PBS, pH 7.4, 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Mouse
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
0.5 mg/mL
Western Blot 0.2-2 μg/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation, Serum
Brain calcium channel II, CACH6calcium channel, R type, alpha-1 polypeptide, CACNL1A6calcium channel, voltage-dependent, alpha 1E subunit, calcium channel, voltage-dependent, R type, alpha 1E subunit, Cav2.3, L type, alpha-1 polypeptide, L type, alpha-1 polypeptide, isoform 6, voltage-dependent calcium channel alpha 1E subunit, voltage-dependent R-type calcium channel subunit alpha-1E, voltage-gated calcium channel alpha 1E subunit, Voltage-gated calcium channel subunit alpha Cav2.3
Immunized with recombinant Mouse CACNA1E (Uniprot#: Q61290; Gly246∽Trp327)
100 μL
Cancer, Endocrinology, Neuroscience, Signal Transduction
777
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.