missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNA1G Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30827-25ug
This item is not returnable.
View return policy
Description
CACNA1G Polyclonal antibody specifically detects CACNA1G in Human samples. It is validated for Western Blot
Specifications
| CACNA1G | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit | |
| Synthetic peptide derived from the rat CACNA1G conjugated to KLH | |
| 25 μg | |
| Neuroscience | |
| 8913 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction