missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNA1I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30828-100ug
This item is not returnable.
View return policy
Description
CACNA1I Polyclonal antibody specifically detects CACNA1I in Human samples. It is validated for Western Blot
Specifications
| CACNA1I | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| Ca(v)3.3, calcium channel, voltage-dependent, T type, alpha 1I subunit, voltage-dependent T-type calcium channel subunit alpha-1I, voltage-dependent, alpha 1I subunit, Voltage-gated calcium channel subunit alpha Cav3.3 | |
| Rabbits were immunized with a synthetic peptide derived from the rat CACNA1I conjugated to KLH | |
| 100 μg | |
| Signal Transduction | |
| 8911 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction