Learn More
Novus Biologicals™ CACNA1I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30828-25ug
 View more versions of this product
Product Code. 30067205
  • 213.00 € / 25µg
Description
CACNA1I Polyclonal antibody specifically detects CACNA1I in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CACNA1I
Polyclonal
Western Blot : 5-10 μg/mL
Ca(v)3.3, calcium channel, voltage-dependent, T type, alpha 1I subunit, voltage-dependent T-type calcium channel subunit alpha-1I, voltage-dependent, alpha 1I subunit, Voltage-gated calcium channel subunit alpha Cav3.3
Rabbits were immunized with a synthetic peptide derived from the rat CACNA1I conjugated to KLH
25 μg
Signal Transduction
8911
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human
Purified