missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNA2D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30829-25ug
This item is not returnable.
View return policy
Description
CACNA2D1 Polyclonal antibody specifically detects CACNA2D1 in Human,Rat samples. It is validated for Western Blot
Specifications
| CACNA2D1 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| CACNA2, CACNL2Adihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit, calcium channel, L type, alpha 2 polypeptide, calcium channel, voltage-dependent, alpha 2/delta subunit 1, CCHL2A, MHS3, voltage-dependent calcium channel subunit alpha-2/delta-1, Voltage-gated calcium channel subunit alpha-2/delta-1 | |
| Synthetic peptide derived from the rat CACNA2D1 conjugated to KLH | |
| 25 μg | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 781 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction