missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CACNB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15587272
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15587272 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15587272 Supplier Invitrogen™ Supplier No. PA546729

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: QDLYQPHRQH TSGLPSANGH DPQDRLLAQD SEHNHSDRNW QRNRPWPKDS Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%.

CACNB3 encodes a regulatory beta subunit of the voltage-dependent calcium channel. Beta subunits are composed of five domains, which contribute to the regulation of surface expression and gating of calcium channels and may also play a role in the regulation of transcription factors and calcium transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNB3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Cacnb3
Gene Accession No. P54284
Gene Alias Beta3; Ca(v)beta3; CAB3; CACH3B; Cacnb3; Cacnlb3; Calcium channel subunit beta 3; calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; calcium voltage-gated channel auxiliary subunit beta 3; cardiac calcium channel beta-subunit CaB3; Cav; CCHB3; voltage-dependent calcium channel beta3A subunit; voltage-dependent L-type calcium channel subunit beta-3
Gene Symbols Cacnb3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CACNB3 (aa 434-483).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 784
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.