missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
166.00 € - 509.00 €
Specifications
| Antigen | CACNG7 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18276582
|
Novus Biologicals
NBP1-30699-0.1MG |
0.1 mg |
166.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18117379
|
Novus Biologicals
NBP1-30699 |
0.5 mg |
509.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNG7 Polyclonal specifically detects CACNG7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CACNG7 | |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| calcium channel, voltage-dependent, gamma subunit 7, Neuronal voltage-gated calcium channel gamma-7 subunit, voltage-dependent calcium channel gamma-7 subunit | |
| CACNG7 | |
| Primary | |
| Appears to be specific for CACNG7. |
| LYOPH | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 59284 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title