missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-4/R-Cadherin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99097-50ul
This item is not returnable.
View return policy
Description
Cadherin-4/R-Cadherin Polyclonal antibody specifically detects Cadherin-4/R-Cadherin in Human samples. It is validated for ELISA
Specifications
| Cadherin-4/R-Cadherin | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
| Produced in rabbits immunized with purified, recombinant Human Cadherin-4/R-Cadherin (Accession#: NP_001785.2; Met1-Ala734). | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 1002 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction