missing translation for 'onlineSavingsMsg'
Learn More

Cadmium standard solution, for AAS, 1 mg/ml Cd in 2 to 5% HNO3

Product Code. 10782811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10782811 500 mL 500mL
10501071 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10782811 Supplier Thermo Scientific Acros Supplier No. 195875000
  • ONLINE EXCLUSIVE PRICE
    105.00 €
    96.70 € / 500mL
    Save 8.30 €
    7% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cadmium standard solution
Name Note 1 mg/ml Cd in 0.5N HNO3
Packaging Plastic bottle
CAS 10022-68-1, 7697-37-2, 7732-18-5
Density 1.0120g/mL
Specific Gravity 1.012
Physical Form Solution
Molecular Formula CdH2N2O6
Concentration 997 to 1003 μg/mL of Cd(2+) in 0.5N HNO3 (20°C)
MDL Number MFCD00149626
Merck Index 15, 1627
Solubility Soluble in water
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O
IUPAC Name cadmium(2+) bis(nitric acid)
Molecular Weight (g/mol) 238.44
PubChem CID 56924536
Quantity 500 mL
ChEBI CHEBI:86156
Formula Weight 308.48
Grade AAS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cadmium atomic absorption standard solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.