missing translation for 'onlineSavingsMsg'
Learn More

Caffeic Acid, TRC

Product Code. 17971955
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971955 1 g 1g
17944785 50 mg 50mg
17902555 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971955 Supplier Toronto Research Chemicals Supplier No. TRCC0800001G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331-39-5
Molecular Formula C9 H8 O4
Molecular Weight (g/mol) 180.16
Synonym Cinnamic acid, 3,4-dihydroxy- (8CI), 3-(3,4-Dihydroxyphenyl)-2-propenoic acid, 3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)propenoic acid, 4-(2-Carboxyethenyl)-1,2-dihydroxybenzene, 4-(2'-Carboxyvinyl)-1,2-dihydroxybenzene, Caffeic acid, DHCA, NSC 57197, NSC 623438
IUPAC Name 3-(3,4-dihydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)C=Cc1ccc(O)c(O)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)
Formula Weight 180.0423
Percent Purity >95%
Chemical Name or Material Caffeic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.